missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue sodium salt, 0.04% w/v aq. soln.

Catalog No. AA38701AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38701AE 100 mL
AA38701AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38701AE Supplier Thermo Scientific Chemicals Supplier No. 038701AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
PubChem CID 102183223
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Odor Odorless
Color Blue
Quantity 100 mL
Physical Form Liquid
IUPAC Name sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
Formula Weight 646.38
Chemical Name or Material Bromothymol Blue sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only