missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18601103
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601103 5 g Amber Glass Bottle
18601104 25 g Amber Glass Bottle
18601105 100 g Amber Glass Bottle
18601106 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601103 Supplier Spectrum Chemical Mfg Cor Supplier No. BR1545GM
Get Up to $500 in Chemicals with Purchase
View all promotions

BR154, 76-59-5, 34722-90-2, C27H28O5SBr2

Spectrum™ Chemical Bromothymol Blue, Reagent, ACS is a pH indicator for weak acids and bases. It is mostly used in applications that require measuring substances that would have a relatively neutral pH (near 7), such as managing the pH of pools and fish tanks. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.38
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
IUPAC Name 3,3-bis[3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC(C)C1=CC(=C(C)C(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(Br)=C1C

Specifications

CAS Max % 100%
Grade ACS
Packaging Amber Glass Bottle
Color Purple/Violet
pH 6.0 to 7.6
Quantity 5 g
Chemical Name or Material Bromothymol Blue