missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue Indicator, 0.04% (w/v), Ricca Chemical
For Glassware Residue Detection
$87.36 - $199.37
Specifications
| CAS | 34722-90-2 , 7732-18-5 |
|---|---|
| Percent Purity | 0.04% W/V |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Color | Dark Blue |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
13754
|
N/A | 120 mL | Natural Poly Bottle |
Each for $87.36
|
|
||||
|
137516
|
N/A | 500 mL | Natural Poly Bottle |
Each for $111.06
|
|
||||
|
137532
|
N/A | 1 L | Natural Poly Bottle |
Each for $199.37
|
|
||||
Specifications
| 34722-90-2 , 7732-18-5 | |
| Indicator | |
| Dark Blue |
| 0.04% W/V | |
| Natural Poly Bottle | |
| 6.70 to 6.90 |