missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Bromothymol Blue, ACS reagent
Supplier: Thermo Scientific™ 403251000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
IndicatorSpecifications
| Bromothymol Blue | |
| 76-59-5 | |
| C27H28Br2O5S | |
| 15, 1455 | |
| NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br | |
| 624.384 | |
| CHEBI:86155 | |
| ACS Reagent | |
| from pH 6.0 (yellow) to pH 7.6 (blue) | |
| 100g |
| Pure | |
| Passes Test | |
| MFCD00005872 | |
| Bromthymol Blue, 3';, 3';';-Dibromothymolsulfonephthalein, BTB | |
| Authentic | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol | |
| 6450 | |
| 624.38 | |
| Glass bottle | |
| Brown-Pink to Purple |
RUO â Research Use Only