missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue

Catalog No. p-7023819
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1774609 10 g
AAA1774618 50 g
AAA1774630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1774609 Supplier Thermo Scientific Chemicals Supplier No. A1774609
Only null left

CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol

Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.

Solubility
Soluble in ethanol, ether and alkalis. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
MDL Number MFCD00005872
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym Bromthymol Blue
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Melting Point 200°C to 202°C
Beilstein 373934
Merck Index 14,1445
Solubility Information Soluble in ethanol,ether and alkalis. Slightly soluble in water.
Formula Weight 624.39
Quantity 10 g
Chemical Name or Material Bromothymol Blue
EINECSNumber 200-971-2
RTECSNumber SJ7450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only