missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromostyrene, 95%, stabilized

Catalog No. AC365800050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC365800050 5 g Glass bottle
AC365800250 25 g Glass bottle
AC365801000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC365800050 Supplier Thermo Scientific Chemicals Supplier No. 365800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-81-7 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-81-7
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
MDL Number MFCD00012229
InChI Key SRXJYTZCORKVNA-UHFFFAOYSA-N
Synonym 1-bromovinyl benzene, alpha-bromostyrene, benzene, 1-bromoethenyl, 1-bromostyrene, styrene, alpha-bromo, 1-phenylvinyl bromide, alpha-bromostyrol russian, styrene, .alpha.-bromo, unii-lf0sj1821n, alpha-bromostyrol
PubChem CID 66828
IUPAC Name 1-bromoethenylbenzene
SMILES BrC(=C)C1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -44°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 67°C to 70°C (4.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(Br)=CH2
Refractive Index 1.5870 to 1.589
Quantity 5 g
Specific Gravity 1.41
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Stabilizer about 0.05% 3, 5-di-tert-butylcatechol
Formula Weight 183.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material α-Bromostyrene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-702-4

RUO – Research Use Only