missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Red, pure

CAS: 2800-80-8 | C19H12Br2O5S | 512.168 g/mol

Supplier:  Thermo Scientific Chemicals 304550050

 View more versions of this product

Catalog No. AC304550050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bromophenol Red
C19H12Br2O5S
5', 5''-Dibromophenolsulfonphthalein
Authentic
2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
76047
512.19
Glass bottle
Brown to Brown/Red
Powder
2800-80-8
MFCD00023014
OYCLSQDXZMROJK-UHFFFAOYSA-N
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br
512.168
5% max. (110°C)
Pure
from pH 5.2 (yellow-red) to pH 7.0 (purple)
5 g

Chemical Identifiers

2800-80-8
512.168
OYCLSQDXZMROJK-UHFFFAOYSA-N
76047
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C=C3)O)Br)C4=CC(=C(C=C4)O)Br
C19H12Br2O5S
MFCD00023014
5', 5''-Dibromophenolsulfonphthalein
2-bromo-4-[3-(3-bromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-538-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only