missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, Sulfone Form, MP Biomedicals
$136.93
Chemical Identifiers
| CAS | 115-39-9 |
|---|---|
| Molecular Formula | C19H10Br4O5S |
| Molecular Weight (g/mol) | 669.96 |
| InChI Key | UDSAIICHUKSCKT-UHFFFAOYSA-N |
| Synonym | 3,3′,5,5′-Tetrabromophenolsulfonphthalein |
| PubChem CID | 8272 |
| ChEBI | CHEBI:59424 |
| IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10112325
|
N/A | 25 g |
Each for $136.93
|
|
|||||
Chemical Identifiers
| 115-39-9 | |
| 669.96 | |
| 3,3′,5,5′-Tetrabromophenolsulfonphthalein | |
| CHEBI:59424 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br |
| C19H10Br4O5S | |
| UDSAIICHUKSCKT-UHFFFAOYSA-N | |
| 8272 | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Specifications
| 115-39-9 | |
| 3,3′,5,5′-Tetrabromophenolsulfonphthalein | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br | |
| 669.96 | |
| CHEBI:59424 | |
| Bromophenol Blue |
| C19H10Br4O5S | |
| UDSAIICHUKSCKT-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 8272 | |
| 670 |