missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Sulfone Form, MP Biomedicals

Catalog No. ICN10112325
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10112325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10112325 Supplier MP Biomedicals Inc Supplier No. 0210112325

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym 3,3′,5,5′-Tetrabromophenolsulfonphthalein
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Formula Weight 670
Quantity 25 g
Chemical Name or Material Bromophenol Blue