missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, Sodium Salt, MP Biomedicals™
Bromophenol Blue, Sodium Salt is a tracking dye.
$58.81 - $58.81
Chemical Identifiers
| Synonym | 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt, 3', 3”, 5' |
|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15250605
|
MP Biomedicals Inc
0215250605 |
5 g |
Each for $58.81
|
|
|||||
Description
- Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.
- It is a tracking dye for alkaline and neutral buffer systems, as well as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.
Chemical Identifiers
| 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt, 3', 3”, 5' |
Specifications
| 62625-28-9 | |
| 3',3”,5',5”-tetrabromophenolsulfonphthalein sodium salt, 3', 3”, 5' | |
| C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] | |
| 691.942 | |
| 692 | |
| 5 g | |
| Bromophenol blue, sodium salt, For ACS analysis |
| C19H9Br4NaO5S | |
| TWKDJMRHSLWRFI-UHFFFAOYSA-M | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
| 3791690 | |
| Green | |
| Crystalline Powder |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.