missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent
Indicator | CAS: 34725-61-6 | C19H9Br4NaO5S | 691.94 g/mol
$61.36 - $585.41
Chemical Identifiers
| CAS | 34725-61-6 |
|---|---|
| Molecular Formula | C19H9Br4NaO5S |
| Molecular Weight (g/mol) | 691.94 |
| MDL Number | MFCD00013793 |
| InChI Key | KGANHHDZDYZJEM-UHFFFAOYNA-M |
| Synonym | 3',3'',5',5''-Tetrabromophenolsulfonephthalein |
| PubChem CID | 3791690 |
| IUPAC Name | sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC403160050
|
Thermo Scientific Chemicals
403160050 |
5 g |
Each for $61.36
|
|
|||||
|
AC403160100
|
Thermo Scientific Chemicals
403160100 |
10 g |
Each for $108.31
Case of 6 Each for $0.00
|
|
|||||
|
AC403160500
|
Thermo Scientific Chemicals
403160500 |
50 g |
Each for $281.58
|
|
|||||
|
AC151351000
|
Thermo Scientific Chemicals
151351000 |
100 g |
Each for $585.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 34725-61-6 | |
| 691.94 | |
| KGANHHDZDYZJEM-UHFFFAOYNA-M | |
| 3791690 |
| C19H9Br4NaO5S | |
| MFCD00013793 | |
| 3',3'',5',5''-Tetrabromophenolsulfonephthalein | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
Specifications
| >300.0°C | |
| C19H9Br4NaO5S | |
| 19, I, 649 | |
| KGANHHDZDYZJEM-UHFFFAOYNA-M | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
| 3791690 | |
| ACS Reagent | |
| 5 g |
| 34725-61-6 | |
| MFCD00013793 | |
| 3',3'',5',5''-Tetrabromophenolsulfonephthalein | |
| C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Br)C3=CC(=C(C(=C3)Br)O)Br)S(=O)(=O)[O-].[Na+] | |
| 691.94 | |
| 691.94 | |
| Glass bottle | |
| Bromophenol Blue, sodium salt, Pure |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Warning
RUO – Research Use Only