Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS reagent
Indicator | CAS: 34725-61-6 | C19H9Br4NaO5S | 691.94 g/mol
Supplier: Thermo Scientific Chemicals 151351000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Bromophenol Blue, sodium salt | |
| Pure | |
| C19H9Br4NaO5S | |
| 19, I, 649 | |
| KGANHHDZDYZJEM-UHFFFAOYNA-M | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate | |
| 3791690 | |
| Pure | |
| 100 g |
| >300.0°C | |
| 34725-61-6 | |
| MFCD00013793 | |
| 3',3'',5',5''-Tetrabromophenolsulfonephthalein | |
| [Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br | |
| 691.94 | |
| 691.94 | |
| Glass bottle |
Chemical Identifiers
| 34725-61-6 | |
| 691.94 | |
| KGANHHDZDYZJEM-UHFFFAOYNA-M | |
| 3791690 |
| C19H9Br4NaO5S | |
| MFCD00013793 | |
| 3',3'',5',5''-Tetrabromophenolsulfonephthalein | |
| sodium;2-[(3,5-dibromo-4-hydroxyphenyl)-(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Warning
RUO – Research Use Only