missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, pure, indicator

Bromophenol Blue, C19H10Br4O5S, CAS Number-115-39-9, 3', 3″, 5', 3',3″,5',5″-Tetrabromophenolsulfonephthalein, BPB, 25g, 19, I, 649, CHEBI:59424, Orange-Pink to Red, 669.96, Pure, UDSAIICHUKSCKT-UHFFFAOYSA-N, Authentic | CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol

Supplier:  Thermo Scientific Chemicals 151341000

 View more versions of this product

Catalog No. AC151341000

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Bromophenol Blue
Indicator
C19H10Br4O5S
19, I, 649
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
UDSAIICHUKSCKT-UHFFFAOYSA-N
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
CHEBI:59424
Pure
1100 min. (592nm, in buffer)
Orange/Pink to Red
Powder
279.0°C
115-39-9
MFCD00005875
15, 1454
Solubility in water: 0.4g/100mL
Authentic
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
8272
669.96
Glass bottle
from pH 3.0 (yellow) to pH 4.6 (blue)
100 g

Chemical Identifiers

115-39-9
669.96
UDSAIICHUKSCKT-UHFFFAOYSA-N
8272
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
C19H10Br4O5S
MFCD00005875
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
CHEBI:59424
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only