missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 1034-49-7
Molecular Formula C19H17Br2P
Molecular Weight (g/mol) 436.14
MDL Number MFCD00011864
InChI Key YFTMLUSIDVFTKU-UHFFFAOYSA-M
Synonym bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
PubChem CID 2733422
IUPAC Name bromomethyl(triphenyl)phosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC321480050
SDS View Documents
Thermo Scientific Chemicals
321480050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC321480250
SDS View Documents
Thermo Scientific Chemicals
321480250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

1034-49-7
436.14
YFTMLUSIDVFTKU-UHFFFAOYSA-M
2733422
C1=CC=C(C=C1)[P+](CBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C19H17Br2P
MFCD00011864
bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
bromomethyl(triphenyl)phosphanium;bromide

Specifications

1034-49-7
100.0
97.5% min. (Argentometry)
C19H17Br2P
MFCD00011864
bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
C1=CC=C(C=C1)[P+](CBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
436.14
436.14
(Bromomethyl)triphenylphosphonium bromide
97.5
Authentic
Glass bottle
BrCH2P(C6H5)3Br
5g
YFTMLUSIDVFTKU-UHFFFAOYSA-M
bromomethyl(triphenyl)phosphanium;bromide
2733422
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-857-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only