missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)trimethylsilane 97.0+%, TCI America™

Catalog No. p-7119334
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18925G 5 g
B189225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18925G Supplier TCI America Supplier No. B18925G

Chemical Identifiers

CAS 18243-41-9
Molecular Formula C4H11BrSi
Molecular Weight (g/mol) 167.12
MDL Number MFCD00000167
InChI Key ACAUYCZBWABOLI-UHFFFAOYSA-N
Synonym (Trimethylsilyl)methyl Bromide
PubChem CID 87523
IUPAC Name (bromomethyl)trimethylsilane
SMILES C[Si](C)(C)CBr

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 g
UN Number 2924
Formula Weight 167.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)trimethylsilane