accessibility menu, dialog, popup

UserName

CH3SiCH3H3CBr

CAS RN 18243-41-9

IUPAC Name: (bromomethyl)trimethylsilane
Synonyms: (bromomethyl)trimethylsilane
Molecular Weight (g/mol): 167.12
Molecular Formula: C4H11BrSi
InChi Key: ACAUYCZBWABOLI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results