missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclopropane, 97%

Catalog No. p-7043522
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0112203 1 g
AAL0112206 5 g
AAL0112214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0112203 Supplier Thermo Scientific Chemicals Supplier No. L0112203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7051-34-5 | C4H7Br | 135.00 g/mol

(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Bromomethyl)cyclopropane is used in the synthesis of 1,4-dienes by iron-catalyzed cross-coupling with alkenyl Grignard reagents. It has been used as a synthetic building block for the introduction of the cyclopropylmethyl group.

Solubility
Not miscible in water.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents.

Chemical Identifiers

CAS 7051-34-5
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00001306
InChI Key AEILLAXRDHDKDY-UHFFFAOYSA-N
Synonym bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
PubChem CID 81503
SMILES BrCC1CC1

Specifications

Density 1.41
Boiling Point 111°C to 112°C
Flash Point 41°C (105°F)
Refractive Index 1.477
Quantity 1 g
UN Number UN1993
Beilstein 605296
Solubility Information Not miscible in water.
IUPAC Name (bromomethyl)cyclopropane
Formula Weight 135
Percent Purity 97%
Chemical Name or Material (Bromomethyl)cyclopropane
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 230-331-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only