missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclopropane, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 221810050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC221810050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7051-34-5
135.00
AEILLAXRDHDKDY-UHFFFAOYSA-N
81503
BrCC1CC1
C4H7Br
MFCD00001306
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
bromomethylcyclopropane

Specifications

(Bromomethyl)cyclopropane
95.0
1.3800g/mL
41°C
95% min. (GC)
C4H7Br
C3H5CH2Br
MFCD00001306
bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
BrCC1CC1
135.00
135
7051-34-5
100.0
101.0°C to 108.0°C
Authentic
Glass bottle
1.4750 to 1.4790
5g
1.38
AEILLAXRDHDKDY-UHFFFAOYSA-N
bromomethylcyclopropane
81503
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-331-8

SDS
Documents

Documents