missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclopropane, 96%, Thermo Scientific™

Catalog No. p-3735741
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221810010 1g Glass bottle
AC221810050 5g Glass bottle
AC221810250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221810010 Supplier Thermo Scientific Chemicals Supplier No. 221810010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7051-34-5
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00001306
InChI Key AEILLAXRDHDKDY-UHFFFAOYSA-N
Synonym bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
PubChem CID 81503
IUPAC Name bromomethylcyclopropane
SMILES BrCC1CC1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 1.3800g/mL
Boiling Point 101.0°C to 108.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4790
Linear Formula C3H5CH2Br
Quantity 1g
Specific Gravity 1.38
Formula Weight 135
Percent Purity 96%
Chemical Name or Material (Bromomethyl)cyclopropane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-331-8

RUO – Research Use Only