missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclohexane 97.0+%, TCI America™

Catalog No. B170825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B170825G 25 g
B1708100G 100 g
B1708500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B170825G Supplier TCI America Supplier No. B170825G

Chemical Identifiers

CAS 2550-36-9
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00001509
InChI Key UUWSLBWDFJMSFP-UHFFFAOYSA-N
Synonym bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane
PubChem CID 137636
IUPAC Name (bromomethyl)cyclohexane
SMILES BrCC1CCCCC1

Specifications

Color Yellow
Boiling Point 94°C
Quantity 25 g
Formula Weight 177.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)cyclohexane