missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclohexane, 98%

Catalog No. AAL1134306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1134306 5 g
AAL1134314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1134306 Supplier Thermo Scientific Chemicals Supplier No. L1134306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2550-36-9 | C7H13Br | 177.09 g/mol

(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 2550-36-9
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00001509
InChI Key UUWSLBWDFJMSFP-UHFFFAOYSA-N
Synonym bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane
PubChem CID 137636
IUPAC Name bromomethylcyclohexane
SMILES BrCC1CCCCC1

Specifications

Density 1.269
Boiling Point 99°C to 100°C (40 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.4935
Quantity 5 g
Beilstein 635738
Solubility Information Insoluble in water.
Formula Weight 177.09
Percent Purity 98%
Chemical Name or Material (Bromomethyl)cyclohexane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only