missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)chlorodimethylsilane 95.0+%, TCI America™

Catalog No. p-7118707
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08475G 5 g
B084725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08475G Supplier TCI America Supplier No. B08475G
Only null left

Chemical Identifiers

CAS 16532-02-8
Molecular Formula C3H8BrClSi
Molecular Weight (g/mol) 187.536
MDL Number MFCD00000503
InChI Key CAURZYXCQQWBJO-UHFFFAOYSA-N
Synonym bromomethyl chlorodimethylsilane, bromomethyldimethylchlorosilane, bromomethyl dimethylchlorosilane, bromomethyl chloro dimethylsilane, bmdmcs, silane, bromomethyl chlorodimethyl, bromomethylchlorodimethylsilane, acmc-209dsc, bromomethyl-dimethyl-chlorosilane, caurzyxcqqwbjo-uhfffaoysa
PubChem CID 85476
IUPAC Name bromomethyl-chloro-dimethylsilane
SMILES C[Si](C)(CBr)Cl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 2986
Formula Weight 187.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Bromomethyl)chlorodimethylsilane