accessibility menu, dialog, popup

UserName

CH3SiCH3ClBr

CAS RN 16532-02-8

IUPAC Name: (bromomethyl)(chloro)dimethylsilane
Synonyms: (bromomethyl)(chloro)dimethylsilane
Molecular Weight (g/mol): 187.54
Molecular Formula: C3H8BrClSi
InChi Key: CAURZYXCQQWBJO-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results