missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromomethyl Acetate 95.0+%, TCI America™
$128.62 - $367.22
Chemical Identifiers
| CAS | 590-97-6 |
|---|---|
| Molecular Formula | C3H5BrO2 |
| Molecular Weight (g/mol) | 152.98 |
| MDL Number | MFCD00000170 |
| InChI Key | NHYXMAKLBXBVEO-UHFFFAOYSA-N |
| Synonym | bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate |
| PubChem CID | 68536 |
| IUPAC Name | bromomethyl acetate |
| SMILES | CC(=O)OCBr |
Chemical Identifiers
| 590-97-6 | |
| 152.98 | |
| NHYXMAKLBXBVEO-UHFFFAOYSA-N | |
| 68536 | |
| CC(=O)OCBr |
| C3H5BrO2 | |
| MFCD00000170 | |
| bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate | |
| bromomethyl acetate |
Specifications
| 590-97-6 | |
| 134°C | |
| MFCD00000170 | |
| 3272 | |
| NHYXMAKLBXBVEO-UHFFFAOYSA-N | |
| bromomethyl acetate | |
| 68536 | |
| ≥95.0% (GC) | |
| Bromomethyl Acetate |
| Yellow | |
| C3H5BrO2 | |
| 1 g | |
| bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate | |
| CC(=O)OCBr | |
| 152.98 | |
| 152.98 | |
| Liquid |
Safety and Handling
RTECSNumber : AF6300200
TSCA : Yes
Recommended Storage : Refrigerator