missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl Acetate 95.0+%, TCI America™

Catalog No. B39071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39071G 1 g
B39075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39071G Supplier TCI America Supplier No. B39071G

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr

Specifications

Color Yellow
Boiling Point 134°C
Quantity 1 g
UN Number 3272
Formula Weight 152.98
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromomethyl Acetate
RTECSNumber AF6300200
TSCA Yes
Recommended Storage Refrigerator