Learn More
Bromomethyl acetate, 95%
CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol
$494.29 - $859.70
Chemical Identifiers
| CAS | 590-97-6 |
|---|---|
| Molecular Formula | C3H5BrO2 |
| Molecular Weight (g/mol) | 152.98 |
| MDL Number | MFCD00000170 |
| InChI Key | NHYXMAKLBXBVEO-UHFFFAOYSA-N |
| Synonym | bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate |
| PubChem CID | 68536 |
| IUPAC Name | bromomethyl acetate |
| SMILES | CC(=O)OCBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5675506
|
Thermo Scientific Chemicals
H5675506 |
5 g |
Each for $494.29
|
|
|||||
|
AAH5675509
|
Thermo Scientific Chemicals
H5675509 |
10 g |
Each for $859.70
|
|
|||||
Description
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.
Solubility
Miscible with chloroform.
Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 590-97-6 | |
| 152.98 | |
| NHYXMAKLBXBVEO-UHFFFAOYSA-N | |
| 68536 | |
| CC(=O)OCBr |
| C3H5BrO2 | |
| MFCD00000170 | |
| bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate | |
| bromomethyl acetate |
Specifications
| 590-97-6 | |
| 130°C to 133°C | |
| C3H5BrO2 | |
| MFCD00000170 | |
| UN3272 | |
| bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate | |
| NHYXMAKLBXBVEO-UHFFFAOYSA-N | |
| bromomethyl acetate | |
| 68536 | |
| 95% |
| 1.56 | |
| 57°C (135°F) | |
| CH3CO2CH2Br | |
| 5 g | |
| Moisture sensitive | |
| Miscible with chloroform. | |
| CC(=O)OCBr | |
| 152.98 | |
| 152.98 | |
| Bromomethyl acetate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 209-696-2
RTECSNumber : AF6300200
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only