missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%

CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol

$836.92 - $836.92

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC399180100
SDS View Documents
Thermo Scientific Chemicals
399180100
10 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

590-97-6
152.98
NHYXMAKLBXBVEO-UHFFFAOYSA-N
68536
CC(=O)OCBr
C3H5BrO2
MFCD00000170
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
bromomethyl acetate

Specifications

590-97-6
1.5600g/mL
57°C
94% min. (GC)
C3H5BrO2
CH3CO2CH2Br
10 g
bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
CC(=O)OCBr
152.98
152.97
Crystalline Powder or Powder
White to Beige
130.0°C to 133.0°C (750.0 mmHg)
Authentic
Glass bottle
1.4470 to 1.4540 (20°C, 589nm)
MFCD00000170
1.56
NHYXMAKLBXBVEO-UHFFFAOYSA-N
bromomethyl acetate
68536
95%
Bromomethyl acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-696-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only