missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl acetate, 95%

Catalog No. AAH5675506
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5675506 5 g
AAH5675509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5675506 Supplier Thermo Scientific Chemicals Supplier No. H5675506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-97-6 | C3H5BrO2 | 152.98 g/mol

Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl acetate is involved in the preparation of optically active cyclohexene antisepsis agents. It acts as reagent in samarium diiodide-mediated conversion of ketones and aldehydes to 1,2-diacetates.

Solubility
Miscible with chloroform.

Notes
Store in cool place. Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 590-97-6
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00000170
InChI Key NHYXMAKLBXBVEO-UHFFFAOYSA-N
Synonym bromomethanol acetate, methanol, bromo-, acetate, acetic acid bromomethyl ester, acetoxymethyl bromide, methanol, 1-bromo-, 1-acetate, brommethylacetat, acetoxymethylbromid, bromo-methyl acetate, acmc-20aljy, 1-bromomethyl acetate
PubChem CID 68536
IUPAC Name bromomethyl acetate
SMILES CC(=O)OCBr

Specifications

Density 1.56
Boiling Point 130°C to 133°C
Flash Point 57°C (135°F)
Linear Formula CH3CO2CH2Br
Quantity 5 g
UN Number UN3272
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform.
Formula Weight 152.98
Percent Purity 95%
Chemical Name or Material Bromomethyl acetate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 209-696-2
RTECSNumber AF6300200
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only