missing translation for 'onlineSavingsMsg'
Learn More

Bromomalonaldehyde, 97%

Catalog No. AAH3103006
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3103006 5 g
AAH3103018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3103006 Supplier Thermo Scientific Chemicals Supplier No. H3103006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2065-75-0 | C3H3BrO2 | 150.959 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2065-75-0
Molecular Formula C3H3BrO2
Molecular Weight (g/mol) 150.959
MDL Number MFCD00459999
InChI Key SURMYNZXHKLDFO-UHFFFAOYSA-N
Synonym 2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
PubChem CID 74945
IUPAC Name 2-bromopropanedial
SMILES C(=O)C(C=O)Br

Specifications

Melting Point ∽145°C (decomposition)
Linear Formula BrCH(CHO)2
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 150.96
Percent Purity 97%
Chemical Name or Material Bromomalonaldehyde
Hazard Category H302-H318-H335
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
EINECSNumber 430-470-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only