accessibility menu, dialog, popup

UserName

CAS RN 2065-75-0

IUPAC Name: 2-bromopropanedial
Synonyms: 2-bromopropanedial
Molecular Weight (g/mol): 150.96
Molecular Formula: C3H3BrO2
InChi Key: SURMYNZXHKLDFO-UHFFFAOYSA-N
SMILES: BrC(C=O)C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results