missing translation for 'onlineSavingsMsg'
Learn More

Bromoform (stabilized with Ethanol) 98.0+%, TCI America™

Catalog No. p-7136235
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T034825G 25 g
T0348500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T034825G Supplier TCI America Supplier No. T034825G

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.731
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br

Specifications

Melting Point 8°C
Color Yellow
Boiling Point 149°C
Quantity 25 g
UN Number 2515
Formula Weight 252.73
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromoform (stabilized with Ethanol)
EINECSNumber (2)-0040
RTECSNumber PB5600000
TSCA Yes