missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 99+%, stabilized

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

$145.83 - $145.83

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC220690250
SDS View Documents
Thermo Scientific Chemicals
220690250
25 mL Glass bottle
Each for $145.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-25-2
252.73
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
CHEBI:38682
C(Br)(Br)Br
CHBr3
DIKBFYAXUHHXCS-UHFFFAOYSA-N
5558
bromoform

Specifications

75-25-2,64-17-5
White
150°C to 151°C
99% min. (excluding stabilizer) (GC)
CHBr3
25 mL
12,74
15, 1430
Solubility in water: slightly soluble
C(Br)(Br)Br
252.73
5558
252.73
Crystalline Powder
8°C
2.8900g/mL
Authentic
Glass bottle
CHBr3
01, 68
2.89
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
DIKBFYAXUHHXCS-UHFFFAOYSA-N
bromoform
1.5 wt% max. ethanol
CHEBI:38682
99+%
Bromoform
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Danger

EINECSNumber : 200-854-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only