missing translation for 'onlineSavingsMsg'
Learn More

Bromoform, 96%, stabilized

CAS: 75-25-2 | CHBr3 | 252.73 g/mol

$401.89 - $401.89

Chemical Identifiers

CAS 75-25-2
Molecular Formula CHBr3
Molecular Weight (g/mol) 252.73
MDL Number MFCD00000128
InChI Key DIKBFYAXUHHXCS-UHFFFAOYSA-N
Synonym tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
PubChem CID 5558
ChEBI CHEBI:38682
IUPAC Name bromoform
SMILES C(Br)(Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158202500
SDS View Documents
Thermo Scientific Chemicals
158202500
250 mL Glass bottle
Each for $401.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-25-2
252.73
DIKBFYAXUHHXCS-UHFFFAOYSA-N
5558
bromoform
CHBr3
MFCD00000128
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
CHEBI:38682
C(Br)(Br)Br

Specifications

75-25-2,64-17-5
Colorless to Orange-Yellow
150.0°C to 151.0°C
95% min. (GC)
CHBr3
MFCD00000128
01,68
2.63
tribromomethane, methane, tribromo, tribrommethan, methenyl tribromide, methyl tribromide, tribrommethaan, tribromometan, bromoforme, bromoformio, rcra waste number u225
DIKBFYAXUHHXCS-UHFFFAOYSA-N
bromoform
3 to 4 wt% ethanol
CHEBI:38682
96%
Bromoform
8.0°C
2.6300g/mL
Authentic
Glass bottle
CHBr3
250 mL
12,74
15,143
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,benzene,chloroform,ether,,petroleum ether,acetone and oils
C(Br)(Br)Br
252.73
5558
252.73
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-854-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only