missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodimethylsulfonium Bromide 98.0+%, TCI America™
Supplier: TCI America B331125G
Specifications
| Bromodimethylsulfonium Bromide | |
| 82°C | |
| MFCD10000904 | |
| 3261 | |
| [Br-].C[S+](C)Br | |
| 221.94 | |
| 221.94 | |
| Crystalline Powder |
| 50450-21-0 | |
| C2H6Br2S | |
| 25 g | |
| RGVBVVVFSXWUIM-UHFFFAOYSA-M | |
| bromodimethylsulfanium bromide | |
| 11481441 | |
| ≥98.0% (T) |
Chemical Identifiers
| 50450-21-0 | |
| 221.94 | |
| RGVBVVVFSXWUIM-UHFFFAOYSA-M | |
| bromodimethylsulfanium bromide |
| C2H6Br2S | |
| MFCD10000904 | |
| 11481441 | |
| [Br-].C[S+](C)Br |
Safety and Handling
TSCA : No
Recommended Storage : Freezer