accessibility menu, dialog, popup

UserName

CAS RN 50450-21-0

CH3SCH3BrBr

CAS RN 50450-21-0

IUPAC Name: bromodimethylsulfanium bromide
Synonyms: bromodimethylsulfanium bromide
Molecular Weight (g/mol): 221.94
Molecular Formula: C2H6Br2S
InChi Key: RGVBVVVFSXWUIM-UHFFFAOYSA-M
SMILES: [Br-].C[S+](C)Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results