missing translation for 'onlineSavingsMsg'
Learn More

(Bromodifluoromethyl)trimethylsilane 98.0+%, TCI America™

Supplier:  TCI America B43255G

 View more versions of this product

Catalog No. B43255G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(Bromodifluoromethyl)trimethylsilane
Colorless
C4H9BrF2Si
5 g
Bromodifluoro(trimethylsilyl)methane
C[Si](C)(C)C(F)(F)Br
203.102
203.10
Liquid
115262-01-6
108°C
MFCD18641931
1993
WDZVWBWAUSUTTO-UHFFFAOYSA-N
[bromo(difluoro)methyl]-trimethylsilane
14639950
≥98.0% (GC)

Chemical Identifiers

115262-01-6
203.102
WDZVWBWAUSUTTO-UHFFFAOYSA-N
14639950
C[Si](C)(C)C(F)(F)Br
C4H9BrF2Si
MFCD18641931
Bromodifluoro(trimethylsilyl)methane
[bromo(difluoro)methyl]-trimethylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions