accessibility menu, dialog, popup

UserName

CH3SiCH3H3CFFBr

CAS RN 115262-01-6

IUPAC Name: (bromodifluoromethyl)trimethylsilane
Synonyms: (bromodifluoromethyl)trimethylsilane
Molecular Weight (g/mol): 203.1
Molecular Formula: C4H9BrF2Si
InChi Key: WDZVWBWAUSUTTO-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(F)(F)Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results