missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodichloroacetic acid, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 71133-14-7 |
| Physical Form | Neat |
| Linear Formula | BrCl2 CCOOH |
| MDL Number | MFCD00871191 |
| Melting Point | 69°C to 72°C |
| Synonym | Bromdichloroacetic acid |
| Type | Organic Pollutant Standard |
| Quantity | 100 mg |
| Formula Weight | 207.84 |
| Show More |