missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclohexane 98.0+%, TCI America™

Catalog No. B058125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058125G 25 g
B0581100G 100 g
B0581500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. B058125G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B058125G

Chemical Identifiers

CAS 108-85-0
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00003819
InChI Key AQNQQHJNRPDOQV-UHFFFAOYSA-N
Synonym cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
PubChem CID 7960
IUPAC Name bromocyclohexane
SMILES BrC1CCCCC1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 163.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclohexane