accessibility menu, dialog, popup

UserName

Br

CAS RN 108-85-0

IUPAC Name: bromocyclohexane
Synonyms: bromocyclohexane
Molecular Weight (g/mol): 163.06
Molecular Formula: C6H11Br
InChi Key: AQNQQHJNRPDOQV-UHFFFAOYSA-N
SMILES: BrC1CCCCC1
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)