missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclohexane, 98%

Catalog No. p-7081568
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1159322 100 g
AAA1159336 500 g
AAA115930E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1159322 Supplier Thermo Scientific Chemicals Supplier No. A1159322
Only null left

CAS: 108-85-0 | C6H11Br | 163.06 g/mol

Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & Light Sensitive. Store away from strong bases.

Chemical Identifiers

CAS 108-85-0
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.06
MDL Number MFCD00003819
InChI Key AQNQQHJNRPDOQV-UHFFFAOYSA-N
Synonym cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
PubChem CID 7960
IUPAC Name bromocyclohexane
SMILES BrC1CCCCC1

Specifications

Melting Point -57°C
Density 1.335
Boiling Point 166°C to 167°C
Flash Point 62°C (143°F)
Refractive Index 1.495
Quantity 100 g
Beilstein 1098373
Sensitivity Light sensitive
Merck Index 14,2730
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 163.06
Percent Purity 98%
Chemical Name or Material Bromocyclohexane
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-622-2
RTECSNumber GU7171700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only