missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclobutane 96.0+%, TCI America™

Catalog No. p-7124599
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07711G 1 g
C07715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07711G Supplier TCI America Supplier No. C07711G
Only null left

Chemical Identifiers

CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
UN Number 1993
Formula Weight 135.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromocyclobutane