missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclobutane, 95%

Catalog No. AAA1281703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1281703 1 g
AAA1281706 5 g
AAA1281714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1281703 Supplier Thermo Scientific Chemicals Supplier No. A1281703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4399-47-7 | C4H7Br | 135.004 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br

Specifications

Density 1.431
Boiling Point 107°C to 108°C
Flash Point 22°C (71°F)
Refractive Index 1.4795
Quantity 1 g
UN Number UN1993
Beilstein 1900415
Sensitivity Light sensitive
Formula Weight 135.01
Percent Purity 95%
Chemical Name or Material Bromocyclobutane
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 224-530-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only