missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple, sodium salt, pure, water soluble, indicator

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

$61.47 - $651.17

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229610050
SDS View Documents
Thermo Scientific Chemicals
229610050
5 g Glass bottle
Each for $61.47
Only null left
 
AC229610100
SDS View Documents
Thermo Scientific Chemicals
229610100
10 g Glass bottle
Each for $104.79
Only null left
 
AC229610250
SDS View Documents
Thermo Scientific Chemicals
229610250
25 g Glass bottle
Each for $192.92
Only null left
 
AC229611000
SDS View Documents
Thermo Scientific Chemicals
229611000
100 g Glass bottle
Each for $651.17
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

62625-30-3
562.204
KVSJYIXUWNOWBD-UGAWPWHASA-M
23665568
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
C21H15Br2NaO5S
MFCD00148896
BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate

Specifications

62625-30-3
MFCD00148896
15, 1394
Solubility in water: Soluble
Authentic
sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
23665568
Pure
900 min. (590nm, 0.01N NaOH)
Brown to Red
Powder
C21H15Br2NaO5S
19, 91
BCP, Bromcresol Purple, sodium salt, 5';, 5';';-Dibromo-o-cresolsulfonephthalein, sodium salt
KVSJYIXUWNOWBD-UGAWPWHASA-M
CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]
562.204
562.2
Glass bottle
from pH 5.2 (yellow) to p 6.8 (purple)
5 g
Bromocresol Purple, sodium salt, Water soluble, Indicator, Pure
Videos
SDS
Documents

Documents

RUO – Research Use Only