missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple sodium salt, 0.04% w/v aq. soln.

Catalog No. AA38700AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38700AE 100 mL
AA38700AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38700AE Supplier Thermo Scientific Chemicals Supplier No. 038700AE

CAS: 62625-30-3 | C21H15Br2NaO5S | 562.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-30-3
Molecular Formula C21H15Br2NaO5S
Molecular Weight (g/mol) 562.204
MDL Number MFCD00148896
InChI Key KVSJYIXUWNOWBD-UGAWPWHASA-M
Synonym Bromocresol Purple, water soluble
PubChem CID 23665568
IUPAC Name sodium;2-[(E)-(3-bromo-4-hydroxy-5-methylphenyl)-(3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
SMILES CC1=CC(=C(C2=CC=CC=C2S(=O)(=O)[O-])C3=CC(=C(C(=C3)Br)O)C)C=C(C1=O)Br.[Na+]

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 562.22
Color Orange
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Bromocresol Purple sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only