missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple Indicator, Reag. Ph. Eur., Honeywell Fluka™
Indicator, Reag. Ph. Eur.
$110.82 - $4516.74
Chemical Identifiers
| CAS | 115-40-2 |
|---|---|
| Molecular Formula | C21H16Br2O5S |
| Molecular Weight (g/mol) | 540.22 |
| MDL Number | MFCD00011681 |
| InChI Key | ABIUHPWEYMSGSR-UHFFFAOYSA-N |
| PubChem CID | 8273 |
| ChEBI | CHEBI:86154 |
| IUPAC Name | 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol |
| SMILES | CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
60046140
|
Honeywell-Fluka
326425G |
5 g | Glass Bottle |
Each for $110.82
|
|
||||
|
60046139
|
Honeywell Chemicals
3264225G |
25 g | Glass Bottle |
Each for $336.50
|
|
||||
|
60046138
|
Honeywell Chemicals
326421KG |
1 kg | Glass Bottle |
Each for $4,516.74
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 115-40-2 | |
| 540.22 | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| CHEBI:86154 | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
| C21H16Br2O5S | |
| MFCD00011681 | |
| 8273 | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol |
Specifications
| 240°C | |
| C21H16Br2O5S | |
| NONH for all modes of transport | |
| ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol | |
| 8273 | |
| 540.22g/mol | |
| Complies for change range | |
| 5 g |
| 115-40-2 | |
| MFCD00011681 | |
| 359618 | |
| CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 | |
| 540.22 | |
| CHEBI:86154 | |
| Glass Bottle | |
| 5.2 to 6.8 (yellow to purple) | |
| Bromocresol Purple, Reag. Ph. Eur. |
Safety and Handling
EINECSNumber : 204-087-8