missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple Indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046140
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046140 5 g Glass Bottle
60046139 25 g Glass Bottle
60046138 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046140 Supplier Solstice Supplier No. 326425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator, Reag. Ph. Eur.

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Melting Point 240°C
Packaging Glass Bottle
Suitability Complies for change range
UN Number NONH for all modes of transport
Beilstein 359618
pH 5.2 to 6.8 (yellow to purple)
Quantity 5 g
Formula Weight 540.22g/mol
Grade Reagent Ph. Eur.
Chemical Name or Material Bromocresol Purple, Reag. Ph. Eur.
EINECSNumber 204-087-8