missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green Sodium Salt 95.0+%, TCI America™
$205.05 - $810.86
Chemical Identifiers
| CAS | 67763-24-0 |
|---|---|
| Molecular Formula | C21H13Br4NaO5S |
| Molecular Weight (g/mol) | 720.00 |
| MDL Number | MFCD00148898 |
| InChI Key | HEFSGAHJDGZCHA-UHFFFAOYNA-M |
| Synonym | Bromocresol Blue Sodium Salt, Sodium Bromocresol Green |
| PubChem CID | 23675901 |
| IUPAC Name | sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-3-methylbenzen-1-olate |
| SMILES | [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C |
Chemical Identifiers
| 67763-24-0 | |
| 720.00 | |
| HEFSGAHJDGZCHA-UHFFFAOYNA-M | |
| 23675901 | |
| [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C |
| C21H13Br4NaO5S | |
| MFCD00148898 | |
| Bromocresol Blue Sodium Salt, Sodium Bromocresol Green | |
| sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-3-methylbenzen-1-olate |
Specifications
| 230°C | |
| C21H13Br4NaO5S | |
| Bromocresol Blue Sodium Salt, Sodium Bromocresol Green | |
| [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C | |
| 720.00 | |
| 720.00 | |
| 1 g | |
| Bromocresol Green Sodium Salt |
| 67763-24-0 | |
| MFCD00148898 | |
| HEFSGAHJDGZCHA-UHFFFAOYNA-M | |
| sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-3-methylbenzen-1-olate | |
| 23675901 | |
| ≥95.0% (HPLC,T) | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes