accessibility menu, dialog, popup

UserName

CAS RN 67763-24-0

NaCH3BrHOBrOSOOBrOBrH3C

CAS RN 67763-24-0

IUPAC Name: sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Synonyms: sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzenolate
Molecular Weight (g/mol): 720
Molecular Formula: C21H13Br4NaO5S
InChi Key: HEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES: [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Color 
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)