missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Sodium Salt 95.0+%, TCI America™

Catalog No. S00415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00415G 5 g
S00411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00415G Supplier TCI America Supplier No. S00415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67763-24-0 | C21H13Br4NaO5S | 720.00 g/mol

Chemical Identifiers

CAS 67763-24-0 , 1007478-52-5
Molecular Formula C21H13Br4NaO5S
Molecular Weight (g/mol) 720.00
MDL Number MFCD00148898
InChI Key HEFSGAHJDGZCHA-UHFFFAOYNA-M
Synonym Bromocresol Blue Sodium Salt, Sodium Bromocresol Green
PubChem CID 23675901
IUPAC Name sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
SMILES [Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C

Specifications

Melting Point 230°C
pH 3.8 to 5.4
Quantity 5 g
Physical Form Crystal/Powder
Formula Weight 720.00
Percent Purity ≥95.0% (HPLC,T)
Grade Laboratory
Chemical Name or Material Bromocresol Green Sodium Salt